Skip to Content
Merck
All Photos(1)

Documents

W507903

Sigma-Aldrich

trans,trans-2,4-Hexadien-1-ol

≥97%

Synonym(s):

Sorbic alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCH2OH
CAS Number:
Molecular Weight:
98.14
Beilstein:
1719722
EC Number:
MDL number:

biological source

synthetic

vapor density

>1 (vs air)

Assay

≥97%

refractive index

n20/D 1.5 (lit.)

bp

80 °C/12 mmHg (lit.)

mp

28-33 °C (lit.)

density

0.871 g/mL at 25 °C (lit.)

SMILES string

[H]/C(C)=C([H])\C([H])=C(/[H])CO

InChI

1S/C6H10O/c1-2-3-4-5-6-7/h2-5,7H,6H2,1H3/b3-2+,5-4+

InChI key

MEIRRNXMZYDVDW-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Fresh, green, herbal

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Bissett et al.
Photodermatology, photoimmunology & photomedicine, 7(2), 63-67 (1990-04-01)
Albino hairless mice (SkH:HR-1) exposed chronically to suberythemal doses of ultraviolet B (UVB) radiation display visible skin wrinkling and tumors. Topical treatment of mice with solutions of conjugated dienes (2,4-hexadien-1-ol and derivatives of it) prior to each UVB radiation exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service