Skip to Content
Merck
All Photos(1)

Key Documents

W400300

Sigma-Aldrich

Methyl (methylthio)acetate

97%

Synonym(s):

Methyl (methylmercapto)acetate, Methyl S-methylthioglycolate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2COOCH3
CAS Number:
Molecular Weight:
120.17
FEMA Number:
4003
Beilstein:
1902431
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.146

biological source

synthetic

Assay

97%

refractive index

n20/D 1.4650 (lit.)

bp

49-51 °C/12 mmHg (lit.)

density

1.11 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

roasted

SMILES string

COC(=O)CSC

InChI

1S/C4H8O2S/c1-6-4(5)3-7-2/h3H2,1-2H3

InChI key

HZYCAKGEXXKCDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service