Skip to Content
Merck
All Photos(1)

Documents

W364606

Sigma-Aldrich

3-Methyl-2-butenal

≥97%

Synonym(s):

3-Methylcrotonaldehyde, 3,3-Dimethylacrolein, 3,3-Dimethylacrylaldehyde, NSC 149164, Prenal, Senecialdehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCHO
CAS Number:
Molecular Weight:
84.12
FEMA Number:
3646
Beilstein:
1734740
EC Number:
Council of Europe no.:
10354
MDL number:
UNSPSC Code:
12164502
Flavis number:
5.124

vapor pressure

7 mmHg ( 20 °C)

Assay

≥97%

refractive index

n20/D 1.462 (lit.)

bp

133-135 °C (lit.)

density

0.878 g/mL at 20 °C
0.872 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)\C=C(\C)C

InChI

1S/C5H8O/c1-5(2)3-4-6/h3-4H,1-2H3

InChI key

SEPQTYODOKLVSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Mild, fusel oil, butter

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service