Documents
SLW1517/06
Pyrex® Media-Lab Bottles, with-out cap and pouring ring, with printed trace code
capacity 250 mL
Synonym(s):
Pyrex, SciLabware 1517/06
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
borosilicate glass bottle
round bottle
white graduation
Joint
Joints threaded neck (GL45)
feature
graduations
without cap
with printed trace code
without pouring ring
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1517/06
capacity
250 mL
diam. × H
70 mm × 138 mm
General description
  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
Legal Information
PYREX is a registered trademark of Corning, Inc.