GF97527213
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1
foil, 10mm disks, 0.008mm thickness, as rolled
Synonym(s):
FE010250
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
foil (disks - as rolled)
manufacturer/tradename
Goodfellow 975-272-13
size × thickness
10 mm × 0.008 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service