Skip to Content
Merck
All Photos(2)

Documents

GF61271018

Carbon

foil, 50mm disks, thickness 0.2mm, flexible graphite, 99.8%

Synonym(s):

Activated charcoal, C 000305, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.8%

form

foil

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 612-710-18

resistivity

1375 μΩ-cm, 20°C (graphite)

disc size × thickness

50 mm × 0.2 mm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Ledbetter et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(1), 85-88 (1987-01-01)
Cluster ions containing 50, 100, or 200 water molecules and one proton are accelerated to over 275 kV and impacted on thin carbon films on which tobacco mosaic virus has been dispersed. After bombardment the films are examined with a
Z oscillation of mean charges of energetic ions emerging from a carbon foil: Correlation with the shell structure of ions.
Shima et al.
Physical review. A, General physics, 39(8), 4316-4318 (1989-04-15)
Christophe A Marquette et al.
Biosensors & bioelectronics, 19(5), 433-439 (2003-11-19)
A multifunctional bio-sensing chip was designed based on the electrochemiluminescent (ECL) detection of enzymatically produced hydrogen peroxide. Six different oxidases specific for choline, glucose, glutamate, lactate, lysine and urate were non-covalently immobilised on imidodiacetic acid chelating beads (glucose oxidase only)
D J McComas et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(21), 9598-9602 (1991-11-01)
Global imaging of the magnetospheric charged particle population can be achieved by remote measurement of the neutral atoms produced when magnetospheric ions undergo charge exchange with cold exospheric neutral atoms. Previously suggested energetic neutral atom imagers were only able to
Investigation of delta -electron emission in collisions of highly charged fast Ne projectiles with carbon-foil targets.
Schiwietz et al.
Physical review. B, Condensed matter, 41(10), 6262-6271 (1990-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service