CLS49954L
Pyrex® wide-mouth graduated Erlenmeyer flask with screw-cap
capacity 4,000 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 4995
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
polypropylene cap
screw top cap
wide-mouth
sterility
non-sterile
feature
cap
closure type Screw cap
packaging
case of 1
manufacturer/tradename
Corning 4995-4L
capacity
4,000 mL
Related Categories
General description
Features a GL 45 size, PP, plug-seal screw-cap on a wide, 29 mm neck opening that accommodates larger volume serological pipettes. Also good for media prep and storage, or as a culture flask when optional membrane cap is used.
Other Notes
Browse all Erlenmeyer flasks
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.