Skip to Content
Merck
All Photos(1)

Key Documents

C77902

Sigma-Aldrich

Cholesteryl hydrocinnamate

97%

Synonym(s):

3β-Hydroxy-5-cholestene 3-hydrocinnamate, 5-Cholesten-3β-ol 3-hydrocinnamate, Cholesteryl phenylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H54O2
CAS Number:
Molecular Weight:
518.81
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid crystal

optical activity

[α]27/D −24°, c = 2 in chloroform

mp

109.5-110.5 °C (lit.)

transition temp

isotropic phase to cholesteric phase 106 °C
cholesteric phase to isotropic phase 110 °C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)CCc5ccccc5

InChI

1S/C36H54O2/c1-25(2)10-9-11-26(3)31-17-18-32-30-16-15-28-24-29(38-34(37)19-14-27-12-7-6-8-13-27)20-22-35(28,4)33(30)21-23-36(31,32)5/h6-8,12-13,15,25-26,29-33H,9-11,14,16-24H2,1-5H3/t26-,29+,30+,31-,32+,33+,35+,36-/m1/s1

InChI key

KPNKAGLPVPTLGB-ZOJFKXTHSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service