Skip to Content
Merck
All Photos(1)

Documents

808571

Sigma-Aldrich

6-(1-Piperazinylmethyl)-2-pyridinecarboxaldehyde bistosylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H31N3O7S2
Molecular Weight:
549.66
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

SMILES string

[H]C(C1=CC=CC(CN2CCNCC2)=N1)=O.O=S(C3=CC=C(C)C=C3)(O)=O.O=S(C4=CC=C(C)C=C4)(O)=O

InChI

1S/C11H15N3O.2C7H8O3S/c15-9-11-3-1-2-10(13-11)8-14-6-4-12-5-7-14;2*1-6-2-4-7(5-3-6)11(8,9)10/h1-3,9,12H,4-8H2;2*2-5H,1H3,(H,8,9,10)

InChI key

QDGVEYQPPPITKT-UHFFFAOYSA-N

Application

The material is used as a reagent for N-terminal modification of proteins. The secondary amine handle readily reacts with commercially available NHS-esters to convert nonspecific bioconjugation reagents to N-terminally specific reagents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service