Skip to Content
Merck
All Photos(1)

Key Documents

798304

Sigma-Aldrich

2-Bromo-3-(2-ethylhexyl)thiophene

97% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19BrS
CAS Number:
Molecular Weight:
275.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97% (GC)

form

liquid

refractive index

n/D 1.523

density

1.201 g/L at 25 °C

SMILES string

BrC1=C(CC(CC)CCCC)C=CS1

InChI

1S/C12H19BrS/c1-3-5-6-10(4-2)9-11-7-8-14-12(11)13/h7-8,10H,3-6,9H2,1-2H3

InChI key

VWIAAXXJIUXUEC-UHFFFAOYSA-N

Application

Organic semiconducting polymer or small molecule precursor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Zhang et al.
Journal of the American Chemical Society, 130(25), 7812-7813 (2008-05-30)
We designed and synthesized the all-conjugated diblock copolymers poly(3-hexylthiophene-block-3-(2-ethylhexyl)thiophene)s (P(3HT-b-3EHT)s) via a modified Grignard metathesis (GRIM), a type of quasi-living polymerization, and studied their microphase-separated structures. The P(3HT-b-3EHT)s synthesized had well-controlled molecular weights and very narrow polydispersity indices (PDIs), which
Novel polythiophene derivatives functionalized with conjugated side-?chain pendants comprising triphenylamine?/carbazole moieties for photovoltaic cell applications
Wang HJ, .
Polym. Chem., 4(3), 506-519 (2013)
Influence of the Ethylhexyl Side-Chain Content on the Open-Circuit Voltage in rr-Poly(3-hexylthiophene-co-3-(2-ethylhexyl)thiophene) Copolymers
Burkhart, Beate; Khlyabich, Petr P.; Thompson, Barry C.
Macromolecules, 45(9), 3740-3748 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service