Skip to Content
Merck
All Photos(1)

Key Documents

79602

Sigma-Aldrich

3,7,11,18,22,26-Hexaazatricyclo[26.2.2.213,16]tetratriaconta-13,15,28,30,31,33-hexaene

puriss., ≥99.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H46N6
CAS Number:
Molecular Weight:
466.71
Beilstein:
3573398
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

puriss.

Assay

≥99.0% (HPLC)

mp

100-104 °C

SMILES string

C1CNCCCNCc2ccc(CNCCCNCCCNCc3ccc(CNC1)cc3)cc2

InChI

1S/C28H46N6/c1-13-29-14-2-18-32-22-27-9-11-28(12-10-27)24-34-20-4-16-30-15-3-19-33-23-26-7-5-25(6-8-26)21-31-17-1/h5-12,29-34H,1-4,13-24H2

InChI key

QSDRINUOZNLAKB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service