Skip to Content
Merck
All Photos(1)

Documents

760021

Sigma-Aldrich

6-Chloro-4-hydroxy-5-methylpyrimidine

96%

Synonym(s):

4-Chloro-6-hydroxy-5-methylpyrimidine, 6-Chloro-5-methyl-4-pyrimidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5ClN2O
CAS Number:
Molecular Weight:
144.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

198-203 °C

SMILES string

Cc1c(O)ncnc1Cl

InChI

1S/C5H5ClN2O/c1-3-4(6)7-2-8-5(3)9/h2H,1H3,(H,7,8,9)

InChI key

XZDABTAOUACVLS-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service