Skip to Content
Merck
All Photos(1)

Key Documents

759643

Sigma-Aldrich

2-Oxa-5-azaspiro[3.4]octane oxalate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO · xC2H2O4
CAS Number:
Molecular Weight:
113.16 (free base basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

solid

mp

85-94 °C

functional group

carboxylic acid
ether

storage temp.

−20°C

SMILES string

OC(C(O)=O)=O.C1CCNC12COC2

InChI

1S/C6H11NO.C2H2O4/c1-2-6(7-3-1)4-8-5-6;3-1(4)2(5)6/h7H,1-5H2;(H,3,4)(H,5,6)

InChI key

JFOZNINEJYPQQK-UHFFFAOYSA-N

Other Notes

molecular weight: 113.16 (free base basis)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service