Skip to Content
Merck
All Photos(1)

Key Documents

722650

Sigma-Aldrich

2,2,4(2,4,4)-Trimethyl-1,6-hexanediamine

99%

Synonym(s):

1,6-Diamino-2,2,4(2,4,4)-trimethylhexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H22N2
CAS Number:
Molecular Weight:
158.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.465 (lit.)

bp

232 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

SMILES string

CC(CN)CC(C)(C)CCN.CC(CCN)CC(C)(C)CN

InChI

1S/2C9H22N2/c1-8(7-11)6-9(2,3)4-5-10;1-8(4-5-10)6-9(2,3)7-11/h2*8H,4-7,10-11H2,1-3H3

InChI key

IXIXPOVPUKTPFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service