Skip to Content
Merck
All Photos(2)

Key Documents

719293

Sigma-Aldrich

(S)-3-(Boc-amino)-2,4-dimethyl-2-pentanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25NO3
CAS Number:
Molecular Weight:
231.33
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]22/D −14.2°, c = 0.5 in chloroform

mp

54-60 °C

functional group

amine
hydroxyl

SMILES string

CC(C)[C@H](NC(=O)OC(C)(C)C)C(C)(C)O

InChI

1S/C12H25NO3/c1-8(2)9(12(6,7)15)13-10(14)16-11(3,4)5/h8-9,15H,1-7H3,(H,13,14)/t9-/m0/s1

InChI key

AWLRFWWWKJXWTQ-VIFPVBQESA-N

Application

(S)-3-(Boc-amino)-2,4-dimethyl-2-pentanol can be used as an intermediate to prepare:
  • Tris(4S-isopropyl-5,5-dimethyl-2-oxazolinyl)phenylborate ligand, applicable in the synthesis of various metal complexes bearing catalyst properties.[1]
  • Hydroxy isocyanides, important building blocks for the synthesis of (hydroxyalkyl)oxazoline ligands.[2]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Straightforward synthesis of chiral hydroxy isocyanides
Bauer M and Kazmaier U
European Journal of Organic Chemistry, 2009(14), 2360-2366 (2009)
Organometallic Complexes of Bulky, Optically Active, C 3-Symmetric Tris (4 S-isopropyl-5, 5-dimethyl-2-oxazolinyl) phenylborate (ToP*)
Xu S, et al.
Organometallics, 34(14), 3508-3519 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service