Skip to Content
Merck
All Photos(1)

Documents

715050

Sigma-Aldrich

Chloro(tricyclohexylphosphine)gold(I)

97%

Synonym(s):

(Tricyclohexylphosphine)gold(I) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H33AuClP
CAS Number:
Molecular Weight:
512.85
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

218-222 °C

SMILES string

Cl[Au].C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

InChI

1S/C18H33P.Au.ClH/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h16-18H,1-15H2;;1H/q;+1;/p-1

InChI key

IBNPRSSHBVYHPU-UHFFFAOYSA-M

Application

Catalyst for:
  • Formal tandem bicyclizations of styrylindoles with alkynyl alkenones
  • Addition of X-H bonds to carbon-carbon multiple bonds
  • Arylation of pyrazine or pyridine with aryl bromides
  • Hydroarylation of alkenes with indoles
  • Cyclization
  • Hydroamination

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service