Skip to Content
Merck
All Photos(1)

Documents

706779

Sigma-Aldrich

1-Allyl-4-(trifluoromethyl)benzene

95%

Synonym(s):

4-(Trifluoromethyl)allylbenzene, 4-Allylbenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9F3
CAS Number:
Molecular Weight:
186.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.449

density

1.112 g/mL at 25 °C

SMILES string

FC(F)(F)c1ccc(CC=C)cc1

InChI

1S/C10H9F3/c1-2-3-8-4-6-9(7-5-8)10(11,12)13/h2,4-7H,1,3H2

InChI key

AEVBWCOLYXWNMX-UHFFFAOYSA-N

Application

Reactant for:
  • Stereoselective preparation of alkenyl nitriles via FeCl2-catalyzed oxidation
  • Regioselective intermolecular allylic C-H alkylation with bis-sulfoxide/Pd(OAc)2 catalyst

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.9 °F

Flash Point(C)

54.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nurhayat Tabanca et al.
Molecules (Basel, Switzerland), 24(13) (2019-07-03)
Ceratitis capitata, the Mediterranean fruit fly, is one of the most serious agricultural pests worldwide responsible for significant reduction in fruit and vegetable yields. Eradication is expensive and often not feasible. Current control methods include the application of conventional insecticides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service