Skip to Content
Merck
All Photos(1)

Documents

657948

Sigma-Aldrich

3,3-Dimethoxypropionitrile

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CHCH2CN
CAS Number:
Molecular Weight:
115.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

Assay

90%

impurities

≤10% cis and trans 3-Methoxypropenenitrile

refractive index

n20/D 1.414 (lit.)

bp

92.5 °C/30 mmHg (lit.)

density

1.026 g/mL at 25 °C (lit.)

SMILES string

COC(CC#N)OC

InChI

1S/C5H9NO2/c1-7-5(8-2)3-4-6/h5H,3H2,1-2H3

InChI key

JYHSJQNYYLGMEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service