Skip to Content
Merck
All Photos(1)

Documents

632902

Sigma-Aldrich

1-(Dimethylamino)-2-nitroethylene

97%

Synonym(s):

N,N-Dimethyl-2-nitroethyleneamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O2
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

103-107 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)\C=C\[N+]([O-])=O

InChI

1S/C4H8N2O2/c1-5(2)3-4-6(7)8/h3-4H,1-2H3/b4-3+

InChI key

JKOVQYWMFZTKMX-ONEGZZNKSA-N

Application

1-(Dimethylamino)-2-nitroethylene undergoes addition-elimination reaction with indoles to form the corresponding 3-(2-nitrovinyl)indoles.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of 2-and 3-Indolylpyrroles via 1, 3-Dipolar Cycloadditions of Munchnones and Nitroalkenes"
Lopchuk.MJ and Gribble.WG
Heterocycles, 82(02), 1617-1617 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service