615757
2-Picolinic-d4 acid
98 atom % D
Synonym(s):
2-Pyridine-d4-carboxylic acid
About This Item
Recommended Products
isotopic purity
98 atom % D
Assay
98% (CP)
form
solid
mp
139-142 °C
mass shift
M+4
SMILES string
[2H]c1nc(c([2H])c([2H])c1[2H])C(O)=O
InChI
1S/C6H5NO2/c8-6(9)5-3-1-2-4-7-5/h1-4H,(H,8,9)/i1D,2D,3D,4D
InChI key
SIOXPEMLGUPBBT-RHQRLBAQSA-N
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service