All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H16O2
CAS Number:
Molecular Weight:
276.33
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
95%
form
solid
mp
80-85 °C (lit.)
λmax
249 nm
SMILES string
CC(=C)C(=O)OCc1c2ccccc2cc3ccccc13
InChI
1S/C19H16O2/c1-13(2)19(20)21-12-18-16-9-5-3-7-14(16)11-15-8-4-6-10-17(15)18/h3-11H,1,12H2,2H3
InChI key
MJYSISMEPNOHEG-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xiaoke Zheng et al.
Talanta, 207, 120290-120290 (2019-10-09)
Human T-lymphotropic virus type II (HTLV-II) is a crucial retrovirus that is closely associated with a variety of human diseases. Herein, an ultrasensitive fluorescent HTLV-II DNA detection strategy was developed for the first time based on magnetic nanoparticles (MNPs) and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service