Skip to Content
Merck
All Photos(1)

Key Documents

568198

Sigma-Aldrich

Bis(trichlorosilyl)methane

97%

Synonym(s):

1,1,1,3,3,3-Hexachloro-1,3-disilapropane, Bis(trichlorosilyl)methane, Methylenebis(trichlorosilane)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3SiCH2SiCl3
CAS Number:
Molecular Weight:
282.92
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.468 (lit.)

bp

179-180 °C (lit.)

density

1.545 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)C[Si](Cl)(Cl)Cl

InChI

1S/CH2Cl6Si2/c2-8(3,4)1-9(5,6)7/h1H2

InChI key

ABDDAHLAEXNYRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service