Skip to Content
Merck
All Photos(1)

Key Documents

565121

Sigma-Aldrich

Ethidium bromide-N,N′-bisacrylamide

97%

Synonym(s):

3,8-Bisacrylamido-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Bisacryloyl ethidium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H24BrN3O2
CAS Number:
Molecular Weight:
502.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

248-252 °C (lit.)

λmax

304 nm
418 nm (2nd)

storage temp.

2-8°C

SMILES string

[Br-].CC[n+]1c(-c2ccccc2)c3cc(NC(=O)C=C)ccc3c4ccc(NC(=O)C=C)cc14

InChI

1S/C27H23N3O2.BrH/c1-4-25(31)28-19-12-14-21-22-15-13-20(29-26(32)5-2)17-24(22)30(6-3)27(23(21)16-19)18-10-8-7-9-11-18;/h4-5,7-17H,1-2,6H2,3H3,(H,28,31);1H

InChI key

VVSGFSHAUFPBFM-UHFFFAOYSA-N

Application

Fluorescent monomer

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Alcinesio et al.
Nature communications, 11(1), 2105-2105 (2020-05-02)
3D-printing networks of droplets connected by interface bilayers are a powerful platform to build synthetic tissues in which functionality relies on precisely ordered structures. However, the structural precision and consistency in assembling these structures is currently limited, which restricts intricate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service