Skip to Content
Merck
All Photos(1)

Key Documents

564036

Sigma-Aldrich

3-Fluoro-4-methoxyphenylboronic acid

≥95%

Synonym(s):

3-Fluoro-4-methoxybenzeneboronic acid, 3-Fluoro-4-methyloxyphenylboronic acid, 4-Methoxy-3-fluorophenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OCH3)B(OH)2
CAS Number:
Molecular Weight:
169.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

mp

206-211 °C (lit.)

SMILES string

COc1ccc(cc1F)B(O)O

InChI

1S/C7H8BFO3/c1-12-7-3-2-5(8(10)11)4-6(7)9/h2-4,10-11H,1H3

InChI key

IILGLPAJXQMKGQ-UHFFFAOYSA-N

General description

Contains varying amounts of anhydride

Application

Reactant for:
  • Preparation of hydroxyphenylnaphthols as 17ß-hydroxysteroid dehydrogenase Type 2 inhibitors
  • Regioselective Suzuki coupling
  • Ruthenium-catalyzed arylation reactions
  • Synthesis of amino-trimethoxyphenyl-aryl thiazoles as microtubule inhibitors and potential antitumors
  • Rhodium catalyzed cyanation
  • Petasis reaction

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service