Recommended Products
Assay
97%
mp
90-94 °C (lit.)
SMILES string
Cc1cc(C)cc(c1)S(Cl)(=O)=O
InChI
1S/C8H9ClO2S/c1-6-3-7(2)5-8(4-6)12(9,10)11/h3-5H,1-2H3
InChI key
LSAGRAXLOLZVKO-UHFFFAOYSA-N
Application
3,5-Dimethylbenzenesulfonyl chloride (dmbSO2Cl) may be used to synthesize hexa-2,4-diyne-1,6-diyl bis(3,5-dimethylbenzenesulfonate) and C2-symmetric ligands, N(SO2R)(CH2Z)2 [where, Z = CH2NH2; R = dmb].
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Structure of hexa-2, 4-diyne-1, 6-diyl bis (3, 5-dimethylbenzenesulfonate).
Acta Crystallographica Section C, Structural Chemistry, 45(11), 1821-1822 (1989)
Inorganic chemistry, 53(2), 1144-1155 (2014-01-10)
Tertiary sulfonamide nitrogen-to-metal bonds of normal length are very rare. We recently discovered such a bond in one class of fac-[Re(CO)3(N(SO2R)(CH2Z)2)](n) complexes (Z = 2-pyridyl) with N(SO2R)dpa ligands derived from di-(2-picolyl)amine (N(H)dpa). fac-[M(CO)3(N(SO2R)(CH2Z)2)](n) agents (M = (186/188)Re, (99m)Tc) could find
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service