Skip to Content
Merck
All Photos(1)

Key Documents

543926

Sigma-Aldrich

Cyclohexanethiol

97%

Synonym(s):

Cyclohexyl mercaptan, Hexahydrothiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11SH
CAS Number:
Molecular Weight:
116.22
Beilstein:
1236342
EC Number:
MDL number:

vapor density

4 (vs air)

vapor pressure

10.3 mmHg ( 37.7 °C)

Assay

97%

availability

available only in EU

refractive index

n20/D 1.493 (lit.)

bp

158-160 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

SC1CCCCC1

InChI

1S/C6H12S/c7-6-4-2-1-3-5-6/h6-7H,1-5H2

InChI key

CMKBCTPCXZNQKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Arkema Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service