All Photos(1)
About This Item
Linear Formula:
Cl2C6H3CH2NH2
CAS Number:
Molecular Weight:
176.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
97%
refractive index
n20/D 1.583 (lit.)
bp
136-140 °C/18 mmHg (lit.)
density
1.321 g/mL at 25 °C (lit.)
SMILES string
NCc1cccc(Cl)c1Cl
InChI
1S/C7H7Cl2N/c8-6-3-1-2-5(4-10)7(6)9/h1-3H,4,10H2
InChI key
JHBVZGONNIVXFJ-UHFFFAOYSA-N
General description
2,3-Dichlorobenzylamine is a halogenated benzylamine derivative. It undergoes oxidation with hydrogen peroxide in the presence of V2O5 to afford N-(2,3-dichlorobenzyl) 2,3-dichlorobenzaldimine.
Application
2,3-Dichlorobenzylamine may be used in the synthesis of 5-(2′,3′-dichlorobenzylamino)uracil and 8-substituted quinolines.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
221.0 °F - closed cup
Flash Point(C)
105 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Novel 5-(N-Alkylaminouracil) Acyclic Nucleosides.
Boncel S, et al.
Synthesis, 2011(04), 603-610 (2011)
J David Becherer et al.
Journal of medicinal chemistry, 58(17), 7021-7056 (2015-08-13)
Starting from the micromolar 8-quinoline carboxamide high-throughput screening hit 1a, a systematic exploration of the structure-activity relationships (SAR) of the 4-, 6-, and 8-substituents of the quinoline ring resulted in the identification of approximately 10-100-fold more potent human CD38 inhibitors.
Guobiao Chu et al.
Organic & biomolecular chemistry, 8(20), 4716-4719 (2010-08-18)
The current syntheses of imines from benzylamines are often performed in organic solvents or under harsh reaction conditions. Clean oxidation of primary benzylamines to imines has been successfully achieved using H(2)O(2) in water at room temperature catalyzed by V(2)O(5). Among
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service