All Photos(1)
About This Item
Linear Formula:
F3C6H2CHO
CAS Number:
Molecular Weight:
160.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
refractive index
n20/D 1.478 (lit.)
bp
165 °C (lit.)
density
1.407 g/mL at 25 °C (lit.)
SMILES string
Fc1cc(F)c(F)c(C=O)c1
InChI
1S/C7H3F3O/c8-5-1-4(3-11)7(10)6(9)2-5/h1-3H
InChI key
RQUNVQGCKIBORD-UHFFFAOYSA-N
General description
Enthalpy of vaporization of 2,3,5-trifluorobenzaldehyde at boiling point (438.15K) is 37.975kjoule/mol.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
136.0 °F - closed cup
Flash Point(C)
57.8 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 535-535 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service