Skip to Content
Merck
All Photos(1)

Documents

517054

Sigma-Aldrich

Barium titanium(IV) tetrahydrofurfuryl oxide solution

in tetrahydrofurfuryl alcohol and ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H54BaO12Ti
CAS Number:
Molecular Weight:
791.94
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

concentration

in tetrahydrofurfuryl alcohol and ethyl alcohol

refractive index

n20/D 1.47

bp

176-183 °C

density

1.16 g/mL at 25 °C

SMILES string

C1COC(C1)CO[Ba]OCC2CCCO2.C3COC(C3)CO[Ti](OCC4CCCO4)(OCC5CCCO5)OCC6CCCO6

InChI

1S/6C5H9O2.Ba.Ti/c6*6-4-5-2-1-3-7-5;;/h6*5H,1-4H2;;/q6*-1;+2;+4

InChI key

CQXMMTRBIPRLRB-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.0 °F - closed cup

Flash Point(C)

22.8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service