Skip to Content
Merck
All Photos(1)

Documents

488151

Sigma-Aldrich

Trimethylphenylphosphonium iodide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5P(CH3)3I
CAS Number:
Molecular Weight:
280.09
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

97%

mp

218 °C (dec.) (lit.)

SMILES string

[I-].C[P+](C)(C)c1ccccc1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service