Skip to Content
Merck
All Photos(1)

Key Documents

478571

Sigma-Aldrich

Oxonole Blue dipotassium salt

Dye content 70 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H16K2N4O12S2
CAS Number:
Molecular Weight:
706.74
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 70%

impurities

5-10% ethanol

mp

250 °C (dec.) (lit.)

λmax

630 nm

SMILES string

[K+].[K+].Oc1c(\C=C\C=C\C=C2/C(=O)N(N=C2C([O-])=O)c3ccc(cc3)S(O)(=O)=O)c(nn1-c4ccc(cc4)S(O)(=O)=O)C([O-])=O

InChI

1S/C25H18N4O12S2.2K/c30-22-18(20(24(32)33)26-28(22)14-6-10-16(11-7-14)42(36,37)38)4-2-1-3-5-19-21(25(34)35)27-29(23(19)31)15-8-12-17(13-9-15)43(39,40)41;;/h1-13,30H,(H,32,33)(H,34,35)(H,36,37,38)(H,39,40,41);;/q;2*+1/p-2/b3-1+,4-2+,19-5-;;

InChI key

JQARXPHCHCHRBV-SAJZWCDJSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service