Skip to Content
Merck
All Photos(1)

Key Documents

472832

Sigma-Aldrich

3,6-Di-O-(tert-butyldimethylsilyl)-D-glucal

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H38O4Si2
CAS Number:
Molecular Weight:
374.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D −44°, c = 0.9 in chloroform

mp

43-45 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)OC[C@H]1OC=C[C@@H](O[Si](C)(C)C(C)(C)C)[C@@H]1O

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service