Skip to Content
Merck
All Photos(2)

Key Documents

457248

Sigma-Aldrich

Poly(vinyltoluene-co-α-methylstyrene)

Synonym(s):

α-Methylstyrene-vinyltoluene copolymer, Methylstyrene-vinyltoluene copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H4CH3)]x[CH2C(CH3)(C6H5)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

refractive index

n20/D 1.58

acid number

<1 mg KOH/g

iodine value

23

transition temp

Tg (DSC) 52 °C (onset)
softening point 95-101 °C (ring and ball)

solubility

hydrocarbons, esters and ketones: soluble
water, alcohols and glycols: insoluble

density

1.05 g/mL at 25 °C (lit.)

SMILES string

CC(=C)c1ccccc1.Cc2ccc(C=C)cc2.Cc3cccc(C=C)c3.Cc4ccccc4C=C

InChI

1S/2C9H10/c1-8(2)9-6-4-3-5-7-9;1-3-9-7-5-4-6-8(9)2/h2*3-7H,1H2,2H3

InChI key

OSBCHXNIHVRZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Adhesive, paint and lacquer additive.

Features and Benefits

Compatible with acrylic, styrene, butadiene and rosin polymers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service