Skip to Content
Merck
All Photos(1)

Key Documents

440310

Sigma-Aldrich

Resorcinol monobenzoate

97%

Synonym(s):

3-Benzoyloxyphenol, 3-Hydroxyphenyl benzoate, m -Hydroxyphenyl benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2C6H4-3-(OH)
CAS Number:
Molecular Weight:
214.22
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

mp

133-135 °C (lit.)

SMILES string

Oc1cccc(OC(=O)c2ccccc2)c1

InChI

1S/C13H10O3/c14-11-7-4-8-12(9-11)16-13(15)10-5-2-1-3-6-10/h1-9,14H

InChI key

GDESWOTWNNGOMW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Ongenae et al.
Dermatology (Basel, Switzerland), 196(4), 470-473 (1998-07-21)
Resorcinol monobenzoate is an ultraviolet light absorber used in cellulose plastics and is known to cause allergic contact dermatitis reactions. It is the most important allergen in spectacle frames but can be a potential allergen in shoes as well. We
Resorcinol monobenzoate: a potential sensitizer in a computer mouse.
A Goossens et al.
Contact dermatitis, 47(4), 235-235 (2002-12-21)
Allergic contact dermatitis to eyeglass frame nosepieces.
G H Telang et al.
Journal of the American Academy of Dermatology, 31(1), 114-115 (1994-07-01)
R C Tung et al.
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society, 9(4), 234-236 (1998-11-11)
One case of allergic contact dermatitis caused by polyvinyl chloride identification bands is reported. Patch tests in this case were positive for three of four bands tested, resorcinol monobenzoate, and benzoyl peroxide. Manufacturers could not confirm the presence of resorcinol
B M Hausen
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society, 12(2), 93-102 (2001-05-31)
In Switzerland, Germany, and Austria, allergic reactions to balsam of Peru (BP) have now made it the third most common contact allergen. A series of 20 single BP constituents (including resorcinol monobenzoate), established in 1995, was used for patch tests

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service