Skip to Content
Merck
All Photos(1)

Key Documents

435554

Sigma-Aldrich

Ethylenediamine tetrakis(ethoxylate-block-propoxylate) tetrol

average Mn ~8,000

Synonym(s):

Tetronic 150R1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2N[(-CH2CH2O-)x[-CH2CH(CH3)O-]yH]2]2
CAS Number:
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

mol wt

average Mn ~8,000

refractive index

n20/D 1.455

viscosity

1,840 cP(25 °C, Brookfield)(lit.)

transition temp

softening point −17 °C
cloud point 20 °C (1 wt. % aqueous solution)

density

1.01 g/mL at 25 °C

HLB

1.0 - 7.0

InChI

1S/C34H72N2O12/c1-27(45-23-31(5)41-15-11-37)19-35(20-28(2)46-24-32(6)42-16-12-38)9-10-36(21-29(3)47-25-33(7)43-17-13-39)22-30(4)48-26-34(8)44-18-14-40/h27-34,37-40H,9-26H2,1-8H3

InChI key

BFKFABWTAFNFID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terminal secondary tetrahydroxy-functional oligomer.

Application

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Features and Benefits

Surface activity is dependent on the EO/PO ratio. Lower foaming, greater defoaming and reduced gelling tendencies relative to Pluronic® surfactants. Terminal secondary hydroxyl groups have lower reactivity and acidity than the primary hydroxyl groups in Pluronic® surfactants.

Legal Information

Pluronic is a registered trademark of BASF

comparable product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>482.0 °F - closed cup

Flash Point(C)

> 250 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service