Skip to Content
Merck
All Photos(1)

Key Documents

379565

Sigma-Aldrich

Lead(II) phthalocyanine

Dye content 80 %

Synonym(s):

Phthalocyanine lead salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16N8Pb
CAS Number:
Molecular Weight:
719.72
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, 80%

reaction suitability

reagent type: catalyst
core: lead

λmax

698 nm

SMILES string

[PbH2]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.Pb/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

WSQYJDCCFQPFJC-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service