Skip to Content
Merck
All Photos(1)

Key Documents

361968

Sigma-Aldrich

Diethylaluminum chloride solution

1.0 M in heptane

Synonym(s):

Chlorodiethylaluminum, Dichlorotetraethyldialuminium, Diethylaluminum monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2AlCl
CAS Number:
Molecular Weight:
120.56
Beilstein:
4123259
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in heptane

density

0.72 g/mL at 25 °C

SMILES string

CC[Al](Cl)CC

InChI

1S/2C2H5.Al.ClH/c2*1-2;;/h2*1H2,2H3;;1H/q;;+1;/p-1

InChI key

YNLAOSYQHBDIKW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service