Recommended Products
Assay
99%
form
solid
mp
158-160 °C (lit.)
SMILES string
Cc1c(F)cccc1C(O)=O
InChI
1S/C8H7FO2/c1-5-6(8(10)11)3-2-4-7(5)9/h2-4H,1H3,(H,10,11)
InChI key
XMKZAIHFVHJGPV-UHFFFAOYSA-N
General description
3-Fluoro-2-methylbenzoic acid is an aryl fluorinated building block.
Application
3-Fluoro-2-methylbenzoic acid was used in the GC-MS detection of metabolites from cell cultures containing 6-fluoro-3-methylphenol.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Applied and environmental microbiology, 59(7), 2229-2238 (1993-07-01)
Anaerobic sewage sludge was used to enrich a methanogenic m-cresol-degrading consortium. 6-Fluoro-3-methylphenol was synthesized and added to subcultures of the consortium with m-cresol. This caused the accumulation of 4-hydroxy-2-methylbenzoic acid. In a separate experiment, the addition of 3-fluorobenzoic acid caused
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service