Skip to Content
Merck
All Photos(1)

Key Documents

331600

Sigma-Aldrich

trans-4-Methyl-1-cyclohexanecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10CO2H
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

109-111 °C (lit.)

functional group

carboxylic acid

SMILES string

C[C@@H]1CC[C@H](CC1)C(O)=O

InChI

1S/C8H14O2/c1-6-2-4-7(5-3-6)8(9)10/h6-7H,2-5H2,1H3,(H,9,10)/t6-,7-

InChI key

QTDXSEZXAPHVBI-LJGSYFOKSA-N

Application

trans-4-Methyl-1-cyclohexanecarboxylic acid may be used in chemical synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guadalupe Montserrat Valdes Labrada et al.
Bioprocess and biosystems engineering, 41(11), 1635-1649 (2018-07-27)
Waters contaminated with naphthenic acids (NAs) and associated tailings are one of the major environmental challenges associated with the processing of oil sands and production of heavy oil. In the current work biodegradation of linear and cyclic naphthenic acids, namely
Wen Wang et al.
Environmental technology, 42(7), 984-1000 (2019-08-06)
Anoxic co-biodegradation of linear and cyclic naphthenic acids (NAs) namely octanoic acid, trans-4-methyl-1-cyclohexane carboxylic acid (trans-4MCHCA), cis- and trans-4-methyl-1-cyclohexane-acetic acids (cis-4MCHAA and trans-4MCHAA) was investigated in denitrifying biofilm reactors. In all evaluated compositions, co-biodegradation of NAs was coupled to denitrification

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service