292451
(R)-(−)-4,4a,5,6,7,8-Hexahydro-4a-methyl-2(3H)-naphthalenone
97%
Synonym(s):
(R)-(−)-10-Methyl-1(9)-octal-2-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
Beilstein:
2046095
MDL number:
UNSPSC Code:
12352000
Assay
97%
optical activity
[α]19/D −208.3°, c = 1 in ethanol
refractive index
n20/D 1.525 (lit.)
bp
155 °C/17 mmHg (lit.)
density
1.018 g/mL at 20 °C (lit.)
SMILES string
C[C@]12CCCCC1=CC(=O)CC2
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service