Skip to Content
Merck
All Photos(1)

Key Documents

286818

Sigma-Aldrich

Neostigmine methyl sulfate

98%

Synonym(s):

3-(N,N-Dimethylcarbamoyloxy)-N,N,N,-trimethylanilinium methyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22N2O6S
CAS Number:
Molecular Weight:
334.39
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

SMILES string

COS([O-])(=O)=O.CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C

InChI

1S/C12H19N2O2.CH4O4S/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;1-5-6(2,3)4/h6-9H,1-5H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

OSZNNLWOYWAHSS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Reversible inhibitor of acetylcholinesterase that is similar to eserine, but does not cross the blood-brain barrier.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsufumi Sato et al.
Masui. The Japanese journal of anesthesiology, 62(1), 19-26 (2013-02-26)
Anticholinesterase, such as neostigmine, was used to be a standard drug at the end of surgery for reversal of nondepolarizing neuromuscular block. Neostigmine decreases the metabolism of acetylcholine (ACh) at the neuromuscular junction and allows its concentration to increase and
G V Cammu et al.
Anaesthesia and intensive care, 40(6), 999-1006 (2012-12-01)
Six years ago, a study performed in our department reported that the incidence of postoperative residual curarisation (PORC) was 39%. The reassessment of neuromuscular monitoring and reversal of neuromuscular block in routine anaesthetic practice is relevant now that sugammadex has
Jakub Fichna et al.
Pharmacological reports : PR, 64(5), 1146-1154 (2012-12-15)
Animal models of visceral pain have gained much attention as an important tool to elucidate the possible mechanisms underlying functional gastrointestinal (GI) disorders. Here we report the development of a new, minimally invasive behavioral model of abdominal pain induced by
Oyun Khookhor et al.
Neuro endocrinology letters, 34(1), 58-61 (2013-03-26)
We previously demonstrated that the direct microinjection of cholinesterase inhibitor (neostigmine) into the hippocampus in rats activated the hypothalamo-pituitary -adrenal axis and increased the level of norepinephrine in the plasma. In the current study we tried to measure the effects
Martina Grosse-Sundrup et al.
BMJ (Clinical research ed.), 345, e6329-e6329 (2012-10-19)
To determine whether use of intermediate acting neuromuscular blocking agents during general anesthesia increases the incidence of postoperative respiratory complications. Prospective, propensity score matched cohort study. General teaching hospital in Boston, Massachusetts, United States, 2006-10. 18,579 surgical patients who received

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service