All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H9NO6
CAS Number:
Molecular Weight:
191.14
Beilstein:
2417114
MDL number:
Pricing and availability is not currently available.
Assay
98%
SMILES string
N[C@@H](CC(C(O)=O)C(O)=O)C(O)=O
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service