Skip to Content
Merck
All Photos(3)

Documents

269220

Sigma-Aldrich

Tetraiodomethane

97%

Synonym(s):

Carbon tetraiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CI4
CAS Number:
Molecular Weight:
519.63
Beilstein:
1733108
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

168 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

IC(I)(I)I

InChI

1S/CI4/c2-1(3,4)5

InChI key

JOHCVVJGGSABQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Nuri Kursunlu et al.
The Analyst, 142(19), 3689-3698 (2017-08-30)
In this study, a novel pillar[5]arene-quinoline (P5-Q) as an organic material is used to fabricate Langmuir-Blodgett (LB) thin films and its organic vapor sensing properties have been investigated. The LB deposition process is characterized by UV-visible spectroscopy, atomic force microscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service