Skip to Content
Merck
All Photos(1)

Documents

252840

Sigma-Aldrich

(R)-(−)-2-Methyl-2,4-pentanediol

99%

Synonym(s):

(−)-Hexylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2C(CH3)2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
6051650
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]22/D −21°, neat

refractive index

n20/D 1.427 (lit.)

bp

197 °C (lit.)

density

0.92 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](O)CC(C)(C)O

InChI

1S/C6H14O2/c1-5(7)4-6(2,3)8/h5,7-8H,4H2,1-3H3/t5-/m1/s1

InChI key

SVTBMSDMJJWYQN-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(-)-2-Methyl-2,4-pentanediol is a chiral crystallizing agent for protein crystallization.

Application

Chiral reagent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An overview on 2-methyl-2, 4-pentanediol in crystallization and in crystals of biological macromolecules.
Anand K, et al.
Acta crystallographica. Section D, Structural biology, 58(10), 1722-1728 (2002)
Crystallization of lysozyme with (R)-,(S)-and (RS)-2-methyl-2, 4-pentanediol.
Stauber M, et al.
Acta crystallographica. Section D, Structural biology, 71(3), 427-441 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service