Skip to Content
Merck
All Photos(1)

Documents

251690

Sigma-Aldrich

Sodium 3-(benzothiazol-2-ylthio)-1-propanesulfonate

Synonym(s):

3-(Benzothiazol-2-ylthio)-1-propanesulfonic acid sodium salt, ZPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10NNaO3S3
CAS Number:
Molecular Weight:
311.38
EC Number:
MDL number:
UNSPSC Code:
12352100

mp

264 °C (dec.)
≥320 °C (dec.)

SMILES string

[Na+].[O-]S(=O)(=O)CCCSc1nc2ccccc2s1

InChI

1S/C10H11NO3S3.Na/c12-17(13,14)7-3-6-15-10-11-8-4-1-2-5-9(8)16-10;/h1-2,4-5H,3,6-7H2,(H,12,13,14);/q;+1/p-1

InChI key

VRKNGZAPJYUNSN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service