Skip to Content
Merck
All Photos(2)

Key Documents

222666

Sigma-Aldrich

3-Fluoro-4-methylaniline

99%

Synonym(s):

3-Fluoro-p-toluidine, 4-Amino-2-fluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)NH2
CAS Number:
Molecular Weight:
125.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.54 (lit.)

mp

30-32 °C (lit.)

density

1.093 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Cc1ccc(N)cc1F

InChI

1S/C7H8FN/c1-5-2-3-6(9)4-7(5)8/h2-4H,9H2,1H3

InChI key

MGRHBBRSAFPBIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preconcentration of aniline derivatives from aqueous solutions using micellar-enhanced ultrafiltration.
Analyst, 118(1), 23-27 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service