Skip to Content
Merck
All Photos(1)

Key Documents

209252

Sigma-Aldrich

5-Chloro-1,3-dimethoxybenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OCH3)2
CAS Number:
Molecular Weight:
172.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

34-36 °C (lit.)

functional group

chloro

SMILES string

COc1cc(Cl)cc(OC)c1

InChI

1S/C8H9ClO2/c1-10-7-3-6(9)4-8(5-7)11-2/h3-5H,1-2H3

InChI key

WQHNWJBSROXROL-UHFFFAOYSA-N

General description

5-Chloro-1,3-dimethoxybenzene participates in Suzuki-Miyaura cross-coupling reactions of aryl- and heteroaryl chlorides with potassium cyclobutyltrifluoroborate[1].

Application

5-Chloro-1,3-dimethoxybenzene was used in the regioselective synthesis of TMC-264, tricyclic structure having chloro-1H-dibenzo[b,d]pyran-4,6-dione skeleton[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The first total synthesis and structural determination of TMC-264.
Tatsuta K, et al.
Tetrahedron Letters, 49(25), 4036-4039 (2008)
Gary A Molander et al.
The Journal of organic chemistry, 73(19), 7481-7485 (2008-09-02)
Suitable conditions were found for the Suzuki-Miyaura cross-coupling reaction of potassium cyclopropyl- and cyclobutyltrifluoroborates with aryl chlorides. Both of these secondary alkyl trifluoroborates coupled in moderate to excellent yield with electron-rich, electron-poor, and hindered aryl chlorides to give a variety

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service