Skip to Content
Merck
All Photos(1)

Key Documents

206822

Sigma-Aldrich

Naphthochrome Green

Dye content 10 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H16Na2O6
CAS Number:
Molecular Weight:
506.41
Colour Index Number:
44530
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 10%

λmax

362 nm

SMILES string

[Na+].[Na+].Oc1c(cc(\C(c2ccccc2)=C3\C=C(C([O-])=O)C(=O)c4ccccc34)c5ccccc15)C([O-])=O

InChI

1S/C29H18O6.2Na/c30-26-19-12-6-4-10-17(19)21(14-23(26)28(32)33)25(16-8-2-1-3-9-16)22-15-24(29(34)35)27(31)20-13-7-5-11-18(20)22;;/h1-15,30H,(H,32,33)(H,34,35);;/q;2*+1/p-2/b25-22-;;

InChI key

YMXSSEMZYBUPOI-PJXKUKQXSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service