Skip to Content
Merck
All Photos(2)

Key Documents

198803

Sigma-Aldrich

Ethyl pipecolinate

98%

Synonym(s):

2-(Ethoxycarbonyl)piperidine, DL-Pipecolic acid ethyl ester, Ethyl 2-piperidinecarboxylate, NSC 49360

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO2
CAS Number:
Molecular Weight:
157.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

216-217 °C (lit.)

density

1.006 g/mL at 25 °C (lit.)

functional group

ester

storage temp.

2-8°C

SMILES string

CCOC(=O)C1CCCCN1

InChI

1S/C8H15NO2/c1-2-11-8(10)7-5-3-4-6-9-7/h7,9H,2-6H2,1H3

InChI key

SZIKRGHFZTYTIT-UHFFFAOYSA-N

Application

Ethyl pipecolinate was used in the synthesis of N-substituted pipecolinates[1].
Reactant for:
Combinatorial chemistry with hydrazones
Swift hydroamination
Synthesis of HCV NS5B polymerase inhibitors
Preparation of selective androgen receptor modulators
Decarbonylative arylation at sp3 carbon centers
Synthesis of quinoline derivatives as selective a2C-adrenoceptor antagonists

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bn MeOH 4 h 93.
Chemical Communications (Cambridge, England), 1065-1065 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service