Skip to Content
Merck
All Photos(1)

Documents

151394

Sigma-Aldrich

2-Methyl-6-nitrobenzoic acid

97%

Synonym(s):

6-Nitro-o-toluic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CO2H
CAS Number:
Molecular Weight:
181.15
Beilstein:
2050804
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

153-157 °C (lit.)

SMILES string

Cc1cccc(c1C(O)=O)[N+]([O-])=O

InChI

1S/C8H7NO4/c1-5-3-2-4-6(9(12)13)7(5)8(10)11/h2-4H,1H3,(H,10,11)

InChI key

CCXSGQZMYLXTOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The vapour pressure of 2-methyl-6-nitrobenzoic acid was studied by Knudsen mass-loss effusion technique.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monte MJ and Hillesheim DM.
The Journal of Chemical Thermodynamics, 33(1), 103-112 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service